Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AV282255.2 | - | 2 - 3 | 143 | 107 | |
| BC019579.1 | - | 1 - 2 | 76 | 107 | ||
| BC048508.1 | - | 4 - 5 | 366 | 107 | ||
| CA465808.1 | - | 2 - 3 | 123 | 107 | ||
More Information
Set Membership: |
|

Zfand4
Genomic Map