Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290338.1 | NP_001277267.1 | 4 - 5 | 651 | 77 | |
| GenBank mRNA | AK089032.1 | - | 4 - 5 | 649 | 77 | |
| AV282255.2 | - | 4 - 5 | 398 | 77 | ||
| BC048508.1 | - | 6 - 7 | 621 | 77 | ||
| CA465808.1 | - | 4 - 5 | 378 | 77 | ||
More Information
Set Membership: |
|

Zfand4
Genomic Map