Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290338.1 | NP_001277267.1 | 5 - 6 | 799 | 75 | |
| NM_001290339.1 | NP_001277268.1 | 3 - 4 | 499 | 75 | ||
| GenBank mRNA | AK012639.1 | - | 3 - 4 | 500 | 75 | |
| AK089032.1 | - | 5 - 6 | 797 | 75 | ||
More Information
Set Membership: |
|

Zfand4
Genomic Map