Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290338.1 | NP_001277267.1 | 7 - 8 | 2019 | 83 | |
| NM_001290339.1 | NP_001277268.1 | 5 - 6 | 1719 | 83 | ||
| GenBank mRNA | AK006693.1 | - | 2 - 3 | 626 | 83 | |
| AK012639.1 | - | 5 - 6 | 1718 | 83 | ||
| AK089032.1 | - | 7 - 8 | 2016 | 83 | ||
| CX223310.1 | - | 2 - 3 | 151 | 83 | ||
More Information
Set Membership: |
|

Zfand4
Genomic Map