Assay Details
| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005032.2 | - | 2 - 3 | 278 | 86 | |
| AK154976.1 | - | 3 - 4 | 470 | 86 | ||
| BC050908.1 | - | 3 - 4 | 453 | 6416955 | 86 | |
| BC053090.1 | - | 2 - 3 | 538 | 5709246 | 86 | |
| BQ770041.1 | - | 2 - 3 | 337 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

March8
Genomic Map