Assay Details
| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009662.2 | NP_033792.1 | 2 - 3 | 500 | 74 | |
| GenBank mRNA | AK089344.1 | - | 2 - 3 | 454 | 74 | |
| AK136868.1 | - | 2 - 3 | 468 | 74 | ||
| AK137481.1 | - | 2 - 3 | 458 | 74 | ||
| AK169938.1 | - | 2 - 3 | 457 | 74 | ||
| AK170016.1 | - | 2 - 3 | 357 | 74 | ||
| AK170434.1 | - | 2 - 3 | 457 | 74 | ||
| AK170879.1 | - | 2 - 3 | 456 | 74 | ||
| AK171191.1 | - | 2 - 3 | 449 | 74 | ||
| AK171413.1 | - | 2 - 3 | 457 | 74 | ||
| BC139102.1 | - | 2 - 3 | 427 | 8862124 | 74 | |
| BC141213.1 | - | 2 - 3 | 427 | 9055825 | 74 | |
| L42198.1 | - | 2 - 3 | 346 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox5
Genomic Map