Assay Details
| Gene Symbol: Alox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009662.2 | NP_033792.1 | 3 - 4 | 588 | 79 | |
| GenBank mRNA | AK089344.1 | - | 3 - 4 | 542 | 79 | |
| AK136868.1 | - | 3 - 4 | 556 | 79 | ||
| AK137481.1 | - | 3 - 4 | 546 | 79 | ||
| AK169938.1 | - | 3 - 4 | 545 | 79 | ||
| AK170016.1 | - | 3 - 4 | 445 | 79 | ||
| AK170434.1 | - | 3 - 4 | 545 | 79 | ||
| AK170879.1 | - | 3 - 4 | 544 | 79 | ||
| AK171191.1 | - | 3 - 4 | 537 | 79 | ||
| AK171413.1 | - | 3 - 4 | 545 | 79 | ||
| BC139102.1 | - | 3 - 4 | 515 | 8862124 | 79 | |
| BC141213.1 | - | 3 - 4 | 515 | 9055825 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alox5
Genomic Map