Assay Details
| Gene Symbol: Ptcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027275.3 | NP_081551.2 | 10 - 11 | 828 | 65 | |
| GenBank mRNA | AK032831.1 | - | 10 - 11 | 812 | 65 | |
| AK146005.1 | - | 10 - 11 | 817 | 65 | ||
| BC060687.1 | - | 1 - 2 | 49 | 65 | ||
| BC084595.1 | - | 1 - 2 | 78 | 65 | ||
| BC115434.2 | - | 10 - 11 | 824 | 40094203 | 65 | |
More Information
Set Membership: |
|

Ptcd3
Genomic Map