Assay Details
| Gene Symbol: Prmt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201371.2 | NP_958759.2 | 5 - 6 | 1043 | 60 | |
| GenBank mRNA | BC060250.1 | - | 5 - 6 | 613 | 5690424 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt8
Genomic Map