Assay Details
| Gene Symbol: Cd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007657.3 | NP_031683.1 | 1 - 2 | 183 | 80 | |
| GenBank mRNA | AK002251.1 | - | 1 - 2 | 189 | 80 | |
| AK012793.1 | - | 1 - 2 | 189 | 80 | ||
| AK151443.1 | - | 1 - 2 | 149 | 80 | ||
| AK151619.1 | - | 1 - 2 | 136 | 80 | ||
| AK170310.1 | - | 1 - 2 | 136 | 80 | ||
| BC070474.1 | - | 1 - 2 | 183 | 30634238 | 80 | |
| L08115.1 | - | 1 - 2 | 121 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cd9
Genomic Map