Assay Details
| Gene Symbol: Cd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007657.3 | NP_031683.1 | 4 - 5 | 458 | 109 | |
| GenBank mRNA | AK002251.1 | - | 4 - 5 | 464 | 109 | |
| AK012793.1 | - | 4 - 5 | 464 | 109 | ||
| AK151443.1 | - | 4 - 5 | 424 | 109 | ||
| AK151619.1 | - | 4 - 5 | 411 | 109 | ||
| AK170310.1 | - | 4 - 5 | 411 | 109 | ||
| BC070474.1 | - | 4 - 5 | 458 | 30634238 | 109 | |
| L08115.1 | - | 4 - 5 | 396 | 109 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cd9
Genomic Map