Assay Details
| Gene Symbol: Tnfrsf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011609.4 | NP_035739.2 | 2 - 3 | 493 | 88 | |
| GenBank mRNA | AK154388.1 | - | 2 - 3 | 463 | 88 | |
| AK159989.1 | - | 2 - 3 | 417 | 88 | ||
| AK211886.1 | - | 2 - 3 | 297 | 88 | ||
| AY541589.1 | - | 2 - 3 | 174 | 88 | ||
| AY541590.1 | - | 2 - 3 | 174 | 88 | ||
| BC004599.1 | - | 2 - 3 | 409 | 3585060 | 88 | |
| BC052675.1 | - | 2 - 3 | 428 | 30061894 | 88 | |
| CK624043.1 | - | 2 - 3 | 493 | 88 | ||
| L26349.1 | - | 2 - 3 | 461 | 88 | ||
| M59377.1 | - | 2 - 3 | 287 | 88 | ||
| M60468.1 | - | 2 - 3 | 386 | 88 | ||
| X57796.1 | - | 2 - 3 | 464 | 88 | ||
| X59238.1 | - | 2 - 3 | 386 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf1a
Genomic Map