Assay Details
| Gene Symbol: Tnfrsf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011609.4 | NP_035739.2 | 8 - 9 | 1073 | 164 | |
| GenBank mRNA | AK154388.1 | - | 8 - 9 | 1043 | 164 | |
| AK159989.1 | - | 8 - 9 | 997 | 164 | ||
| AY541589.1 | - | 8 - 9 | 754 | 164 | ||
| AY541590.1 | - | 8 - 9 | 754 | 164 | ||
| BC004599.1 | - | 8 - 9 | 989 | 3585060 | 164 | |
| BC052675.1 | - | 8 - 9 | 1008 | 30061894 | 164 | |
| L26349.1 | - | 8 - 9 | 1041 | 164 | ||
| M59377.1 | - | 8 - 9 | 867 | 164 | ||
| M60468.1 | - | 8 - 9 | 966 | 164 | ||
| X57796.1 | - | 8 - 9 | 1044 | 164 | ||
| X59238.1 | - | 8 - 9 | 966 | 164 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf1a
Genomic Map