Assay Details
| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022879.2 | NP_075017.2 | 5 - 6 | 400 | 84 | |
| GenBank mRNA | AJ348866.1 | - | 4 - 5 | 310 | 84 | |
| AK146716.1 | - | 5 - 6 | 401 | 84 | ||
| BC061100.1 | - | 5 - 6 | 396 | 30300846 | 84 | |
| S70785.1 | - | 4 - 5 | 345 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myl7
Genomic Map