Assay Details
| Gene Symbol: Sumf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145937.3 | NP_666049.2 | 6 - 7 | 865 | 58 | |
| GenBank mRNA | AK151874.1 | - | 6 - 7 | 866 | 58 | |
| AK159192.1 | - | 6 - 7 | 854 | 58 | ||
| AK160917.1 | - | 6 - 7 | 847 | 58 | ||
| AK161203.1 | - | 6 - 7 | 858 | 58 | ||
| AK216554.1 | - | 2 - 3 | 183 | 58 | ||
| BC026981.1 | - | 6 - 7 | 838 | 5365580 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sumf1
Genomic Map