Assay Details
| Gene Symbol: Ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025358.3 | NP_079634.2 | 6 - 7 | 682 | 64 | |
| GenBank mRNA | AK003137.1 | - | 6 - 7 | 664 | 64 | |
| AK003214.1 | - | 6 - 7 | 655 | 64 | ||
| AK012366.1 | - | 6 - 7 | 662 | 64 | ||
| BC005760.1 | - | 5 - 6 | 563 | 64 | ||
| BC058378.1 | - | 6 - 7 | 650 | 6820787 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufa9
Genomic Map