Assay Details
| Gene Symbol: Tigar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177003.5 | NP_795977.1 | 2 - 3 | 154 | 83 | |
| GenBank mRNA | AK036082.1 | - | 2 - 3 | 145 | 83 | |
| AK078900.1 | - | 2 - 3 | 145 | 83 | ||
| AK145896.1 | - | 2 - 3 | 139 | 83 | ||
| AK163290.1 | - | 2 - 3 | 130 | 83 | ||
| AK165614.1 | - | 2 - 3 | 155 | 83 | ||
| BC147705.1 | - | 2 - 3 | 144 | 9087464 | 83 | |
| BC147718.1 | - | 2 - 3 | 144 | 9087477 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tigar
Genomic Map