Assay Details
| Gene Symbol: Tigar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177003.5 | NP_795977.1 | 3 - 4 | 273 | 81 | |
| GenBank mRNA | AK036082.1 | - | 3 - 4 | 264 | 81 | |
| AK078900.1 | - | 3 - 4 | 264 | 81 | ||
| AK145896.1 | - | 3 - 4 | 258 | 81 | ||
| AK163290.1 | - | 3 - 4 | 249 | 81 | ||
| AK165614.1 | - | 3 - 4 | 274 | 81 | ||
| BC147705.1 | - | 3 - 4 | 263 | 9087464 | 81 | |
| BC147718.1 | - | 3 - 4 | 263 | 9087477 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tigar
Genomic Map