Assay Details
| Gene Symbol: Atp6v1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007510.2 | NP_031536.2 | 4 - 5 | 336 | 80 | |
| GenBank mRNA | AK010603.1 | - | 4 - 5 | 353 | 80 | |
| AK139791.1 | - | 4 - 5 | 353 | 80 | ||
| AK146162.1 | - | 4 - 5 | 350 | 80 | ||
| AK149356.1 | - | 4 - 5 | 372 | 80 | ||
| AK167644.1 | - | 4 - 5 | 295 | 80 | ||
| AK169854.1 | - | 4 - 5 | 350 | 80 | ||
| AK186366.1 | - | 4 - 5 | 288 | 80 | ||
| AK189499.1 | - | 4 - 5 | 285 | 80 | ||
| AK189578.1 | - | 4 - 5 | 288 | 80 | ||
| BC003421.1 | - | 4 - 5 | 336 | 2598896 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1e1
Genomic Map