Assay Details
| Gene Symbol: Bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153516.2 | NP_705736.1 | 2 - 3 | 289 | 158 | |
| GenBank mRNA | AK047223.1 | - | 2 - 3 | 291 | 158 | |
| AK088417.1 | - | 2 - 3 | 304 | 158 | ||
| AK132774.1 | - | 2 - 3 | 298 | 158 | ||
| AK143457.1 | - | 2 - 3 | 302 | 158 | ||
| AK148007.1 | - | 2 - 3 | 308 | 158 | ||
| AK165068.1 | - | 2 - 3 | 282 | 158 | ||
| AK168833.1 | - | 2 - 3 | 279 | 158 | ||
| AK171256.1 | - | 2 - 3 | 309 | 158 | ||
| BC027307.1 | - | 2 - 3 | 179 | 4014844 | 158 | |
| BC029016.1 | - | 2 - 3 | 289 | 5358785 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl2l13
Genomic Map