Assay Details
| Gene Symbol: Bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153516.2 | NP_705736.1 | 3 - 4 | 397 | 126 | |
| GenBank mRNA | AK031018.1 | - | 3 - 4 | 346 | 126 | |
| AK047223.1 | - | 3 - 4 | 399 | 126 | ||
| AK088417.1 | - | 3 - 4 | 412 | 126 | ||
| AK132774.1 | - | 3 - 4 | 406 | 126 | ||
| AK143457.1 | - | 3 - 4 | 410 | 126 | ||
| AK148007.1 | - | 3 - 4 | 416 | 126 | ||
| AK168833.1 | - | 3 - 4 | 387 | 126 | ||
| AK171256.1 | - | 3 - 4 | 417 | 126 | ||
| AK178476.1 | - | 2 - 3 | 171 | 127 | ||
| AK178590.1 | - | 2 - 3 | 170 | 126 | ||
| AK186867.1 | - | 2 - 3 | 170 | 126 | ||
| AK187777.1 | - | 2 - 3 | 170 | 126 | ||
| BC027307.1 | - | 3 - 4 | 287 | 4014844 | 126 | |
| BC029016.1 | - | 3 - 4 | 397 | 5358785 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl2l13
Genomic Map