Assay Details
| Gene Symbol: Bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153516.2 | NP_705736.1 | 4 - 5 | 557 | 69 | |
| GenBank mRNA | AK031018.1 | - | 4 - 5 | 506 | 69 | |
| AK047223.1 | - | 4 - 5 | 559 | 69 | ||
| AK088417.1 | - | 4 - 5 | 572 | 69 | ||
| AK132774.1 | - | 4 - 5 | 566 | 69 | ||
| AK143457.1 | - | 4 - 5 | 570 | 69 | ||
| AK148007.1 | - | 4 - 5 | 576 | 69 | ||
| AK165068.1 | - | 4 - 5 | 550 | 69 | ||
| AK168833.1 | - | 4 - 5 | 547 | 69 | ||
| AK179659.1 | - | 3 - 4 | 329 | 69 | ||
| BC027307.1 | - | 4 - 5 | 447 | 4014844 | 69 | |
| BC029016.1 | - | 4 - 5 | 557 | 5358785 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcl2l13
Genomic Map