Assay Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 12 - 13 | 1749 | 90 | |
| GenBank mRNA | AK143510.1 | - | 12 - 13 | 1713 | 90 | |
| AK185760.1 | - | 2 - 3 | 260 | 90 | ||
| BC015245.1 | - | 12 - 13 | 1686 | 4242193 | 90 | |
| L03292.2 | - | 12 - 13 | 1721 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a6
Genomic Map