Assay Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 13 - 14 | 1847 | 79 | |
| GenBank mRNA | AK143510.1 | - | 13 - 14 | 1811 | 79 | |
| AK185760.1 | - | 3 - 4 | 359 | 80 | ||
| BC015245.1 | - | 13 - 14 | 1784 | 4242193 | 79 | |
| L03292.2 | - | 13 - 14 | 1819 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a6
Genomic Map