Assay Details
| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028766.2 | NP_083042.1 | 8 - 9 | 837 | 80 | |
| GenBank mRNA | AK004778.1 | - | 8 - 9 | 838 | 80 | |
| AK037165.1 | - | 8 - 9 | 808 | 80 | ||
| AK050173.1 | - | 8 - 9 | 876 | 80 | ||
| AK151693.1 | - | 8 - 9 | 830 | 80 | ||
| AK151766.1 | - | 8 - 9 | 855 | 80 | ||
| AK152078.1 | - | 8 - 9 | 830 | 80 | ||
| AK168559.1 | - | 8 - 9 | 845 | 80 | ||
| BC024933.1 | - | 8 - 9 | 824 | 3990353 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem43
Genomic Map