Assay Details
| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025823.4 | NP_080099.1 | 4 - 5 | 771 | 56 | |
| GenBank mRNA | AK004799.2 | - | 4 - 5 | 726 | 56 | |
| AK004840.1 | - | 4 - 5 | 726 | 56 | ||
| AK031585.1 | - | 4 - 5 | 727 | 56 | ||
| AK033373.1 | - | 4 - 5 | 727 | 56 | ||
| AK049412.1 | - | 4 - 5 | 724 | 56 | ||
| AK134528.1 | - | 4 - 5 | 772 | 56 | ||
| AK145366.1 | - | 4 - 5 | 731 | 56 | ||
| AK147190.1 | - | 4 - 5 | 739 | 56 | ||
| AK159981.1 | - | 4 - 5 | 727 | 56 | ||
| AK161616.1 | - | 4 - 5 | 727 | 56 | ||
| AK169209.1 | - | 4 - 5 | 727 | 56 | ||
| AK173058.1 | - | 4 - 5 | 693 | 56 | ||
| BC006056.1 | - | 2 - 3 | 382 | 56 | ||
| BC028308.1 | - | 4 - 5 | 741 | 3486530 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyox1
Genomic Map