Assay Details
| Gene Symbol: Add2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271857.1 | NP_001258786.1 | 9 - 10 | 1356 | 75 | |
| NM_001271858.1 | NP_001258787.1 | 9 - 10 | 1353 | 75 | ||
| NM_001271859.1 | NP_001258788.1 | 9 - 10 | 1154 | 75 | ||
| NM_001271860.1 | NP_001258789.1 | 9 - 10 | 1356 | 75 | ||
| NM_001271861.1 | NP_001258790.1 | 9 - 10 | 1157 | 75 | ||
| NM_013458.5 | NP_038486.2 | 9 - 10 | 1157 | 75 | ||
| GenBank mRNA | AF100422.1 | - | 8 - 9 | 1032 | 75 | |
| AF100423.1 | - | 8 - 9 | 1032 | 75 | ||
| AF189769.1 | - | 7 - 8 | 954 | 75 | ||
| AK014496.1 | - | 9 - 10 | 1122 | 75 | ||
| AK028425.1 | - | 9 - 10 | 1128 | 75 | ||
| AK029488.1 | - | 9 - 10 | 1307 | 75 | ||
| AK147544.1 | - | 9 - 10 | 1229 | 75 | ||
| AK156954.1 | - | 9 - 10 | 1146 | 75 | ||
| BC046783.1 | - | 9 - 10 | 1290 | 6401200 | 75 | |
| BC053032.1 | - | 9 - 10 | 1290 | 6401200 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Add2
Genomic Map