Assay Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040106.2 | NP_001035195.1 | 15 - 16 | 2796 | 89 | |
| NM_177762.6 | NP_808430.2 | 14 - 15 | 2553 | 89 | ||
| GenBank mRNA | AK147363.1 | - | 15 - 16 | 2797 | 89 | |
| BC043125.1 | - | 4 - 5 | 602 | 89 | ||
| BC141176.1 | - | 15 - 16 | 2552 | 9055788 | 89 | |
More Information
Set Membership: |
|

Aak1
Genomic Map