Assay Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243741.1 | NP_001230670.1 | 2 - 3 | 862 | 93 | |
| NM_001243742.1 | NP_001230671.1 | 2 - 3 | 398 | 93 | ||
| NM_146170.4 | NP_666282.3 | 2 - 3 | 339 | 93 | ||
| GenBank mRNA | AK047127.1 | - | 2 - 3 | 340 | 93 | |
| AK077101.1 | - | 165 | 93 | |||
| AK077503.1 | - | 2 - 3 | 749 | 93 | ||
| AK170524.1 | - | 2 - 3 | 318 | 93 | ||
| AK171511.1 | - | 2 - 3 | 318 | 93 | ||
| BC025632.1 | - | 2 - 3 | 306 | 5321669 | 93 | |
| BC026926.1 | - | 2 - 3 | 409 | 3586097 | 93 | |
| BC031913.1 | - | 2 - 3 | 352 | 5376246 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047127.1, AK077503.1, AK170524.1, AK171511.1, BC025632.1, BC026926.1, BC031913.1, NM_001243741.1, NM_001243742.1, NM_146170.4 transcripts.
Set Membership: |
|

C87436
Genomic Map