Assay Details
| Gene Symbol: Mcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008564.2 | NP_032590.2 | 11 - 12 | 2035 | 89 | |
| GenBank mRNA | AF004105.1 | - | 10 - 11 | 1925 | 89 | |
| AK037751.1 | - | 11 - 12 | 1960 | 89 | ||
| AK077801.1 | - | 11 - 12 | 1942 | 89 | ||
| AK088156.1 | - | 11 - 12 | 1948 | 89 | ||
| AK088378.1 | - | 11 - 12 | 1935 | 89 | ||
| AK129039.1 | - | 11 - 12 | 1916 | 89 | ||
| AK145176.1 | - | 11 - 12 | 1947 | 89 | ||
| AK146187.1 | - | 11 - 12 | 1950 | 89 | ||
| AK146375.1 | - | 11 - 12 | 1959 | 89 | ||
| BC055318.1 | - | 11 - 12 | 1944 | 5696348 | 89 | |
| D86725.1 | - | 11 - 12 | 1955 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm2
Genomic Map