Assay Details
| Gene Symbol: Mcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008564.2 | NP_032590.2 | 3 - 4 | 543 | 107 | |
| GenBank mRNA | AF004105.1 | - | 2 - 3 | 107 | ||
| AK037751.1 | - | 3 - 4 | 467 | 107 | ||
| AK088156.1 | - | 3 - 4 | 456 | 107 | ||
| AK088378.1 | - | 3 - 4 | 444 | 108 | ||
| AK129039.1 | - | 3 - 4 | 424 | 107 | ||
| AK145176.1 | - | 3 - 4 | 454 | 107 | ||
| AK146187.1 | - | 3 - 4 | 458 | 107 | ||
| AK146375.1 | - | 3 - 4 | 467 | 107 | ||
| BC055318.1 | - | 3 - 4 | 452 | 5696348 | 107 | |
| D86725.1 | - | 3 - 4 | 463 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm2
Genomic Map