Assay Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271758.1 | NP_001258687.1 | 1 - 2 | 118 | 58 | |
| GenBank mRNA | BC010775.1 | - | 1 - 2 | 118 | 4211437 | 58 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC010775.1, NM_001271758.1 transcripts.
Set Membership: |
|

Wnt5b
Genomic Map