Assay Details
| Gene Symbol: Iqsec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033354.3 | NP_001028526.1 | 8 - 9 | 2711 | 96 | |
| NM_001289685.1 | NP_001276614.1 | 7 - 8 | 2440 | 96 | ||
| NR_110358.1 | 7 - 8 | 2440 | 96 | |||
| GenBank mRNA | AK122446.1 | - | 7 - 8 | 2427 | 96 | |
| AK147351.1 | - | 7 - 8 | 2386 | 96 | ||
| AK169437.1 | - | 8 - 9 | 2711 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iqsec3
Genomic Map