Assay Details
| Gene Symbol: Vax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011912.3 | NP_036042.1 | 1 - 2 | 281 | 76 | |
| GenBank mRNA | AB221556.1 | - | 1 - 2 | 252 | 76 | |
| AF028715.1 | - | 1 - 2 | 255 | 76 | ||
| BC090635.1 | - | 1 - 2 | 278 | 30942578 | 76 | |
| BC116389.1 | - | 1 - 2 | 273 | 40101824 | 76 | |
| BC116390.1 | - | 1 - 2 | 273 | 40101825 | 76 | |
| Y17792.1 | - | 1 - 2 | 281 | 76 | ||
More Information
Set Membership: |
|

Vax2
Genomic Map