| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600019880.mm.1 |
NM_023790.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019880.mm.1 |
NM_023790.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019880.mm.1 |
TC0600002562.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019884.mm.1 |
NM_023790.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019884.mm.1 |
NM_023790.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019884.mm.1 |
TC0600002562.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019883.mm.1 |
NM_023790.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019883.mm.1 |
NM_023790.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019883.mm.1 |
TC0600002562.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019888.mm.1 |
NM_023790.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019888.mm.1 |
NM_023790.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019888.mm.1 |
TC0600002562.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019881.mm.1 |
NM_023790.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019881.mm.1 |
NM_023790.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019881.mm.1 |
TC0600002562.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600019886.mm.1 |
NM_023790.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019886.mm.1 |
NM_023790.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600019886.mm.1 |
TC0600002562.mm.1 |
MTA TC |