Assay Details
| Gene Symbol: Wdr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023790.2 | NP_076279.1 | 7 - 8 | 843 | 61 | |
| GenBank mRNA | AF053628.1 | - | 7 - 8 | 831 | 61 | |
| AK006536.1 | - | 7 - 8 | 743 | 61 | ||
| AK154345.1 | - | 7 - 8 | 834 | 61 | ||
| BC083332.1 | - | 8 - 9 | 748 | 6819838 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr54
Genomic Map