Assay Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277330.1 | NP_001264259.1 | 2 - 3 | 258 | 91 | |
| NM_001277805.1 | NP_001264734.1 | 2 - 3 | 256 | 91 | ||
| NM_008995.2 | NP_033021.2 | 2 - 3 | 219 | 91 | ||
| NM_175933.2 | NP_787947.1 | 2 - 3 | 256 | 91 | ||
| GenBank mRNA | AJ416473.1 | - | 2 - 3 | 238 | 91 | |
| AK088886.1 | - | 2 - 3 | 260 | 91 | ||
| AK145111.1 | - | 2 - 3 | 192 | 91 | ||
| AK145361.1 | - | 2 - 3 | 193 | 91 | ||
| AK161470.1 | - | 2 - 3 | 253 | 91 | ||
| BC029748.1 | - | 2 - 3 | 226 | 2599159 | 91 | |
| BY122705.1 | - | 2 - 3 | 257 | 91 | ||
| Z97018.1 | - | 1 - 2 | 144 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex5
Genomic Map