Assay Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277330.1 | NP_001264259.1 | 3 - 4 | 297 | 63 | |
| NM_001277805.1 | NP_001264734.1 | 3 - 4 | 295 | 63 | ||
| NM_008995.2 | NP_033021.2 | 3 - 4 | 258 | 63 | ||
| NM_175933.2 | NP_787947.1 | 3 - 4 | 295 | 63 | ||
| GenBank mRNA | AJ416473.1 | - | 3 - 4 | 277 | 63 | |
| AK088886.1 | - | 3 - 4 | 299 | 63 | ||
| AK145111.1 | - | 3 - 4 | 231 | 63 | ||
| AK145361.1 | - | 3 - 4 | 232 | 63 | ||
| AK161470.1 | - | 3 - 4 | 292 | 63 | ||
| BC029748.1 | - | 3 - 4 | 265 | 2599159 | 63 | |
| BY122705.1 | - | 3 - 4 | 296 | 63 | ||
| Z97018.1 | - | 2 - 3 | 183 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex5
Genomic Map