Assay Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277330.1 | NP_001264259.1 | 6 - 7 | 667 | 105 | |
| NM_001277805.1 | NP_001264734.1 | 6 - 7 | 665 | 105 | ||
| NM_008995.2 | NP_033021.2 | 6 - 7 | 628 | 105 | ||
| NM_175933.2 | NP_787947.1 | 6 - 7 | 665 | 105 | ||
| GenBank mRNA | AJ416473.1 | - | 6 - 7 | 647 | 105 | |
| AK088886.1 | - | 6 - 7 | 669 | 105 | ||
| AK145111.1 | - | 6 - 7 | 601 | 105 | ||
| AK145361.1 | - | 6 - 7 | 602 | 105 | ||
| AK161470.1 | - | 6 - 7 | 662 | 105 | ||
| BC029748.1 | - | 6 - 7 | 635 | 2599159 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pex5
Genomic Map