Assay Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277805.1 | NP_001264734.1 | 8 - 9 | 865 | 88 | |
| NM_008995.2 | NP_033021.2 | 8 - 9 | 828 | 88 | ||
| GenBank mRNA | AK145111.1 | - | 8 - 9 | 801 | 88 | |
| BC029748.1 | - | 8 - 9 | 835 | 2599159 | 88 | |
| Z97018.1 | - | 7 - 8 | 753 | 88 | ||
More Information
Set Membership: |
|

Pex5
Genomic Map