Assay Details
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277330.1 | NP_001264259.1 | 8 - 9 | 857 | 69 | |
| NM_001277805.1 | NP_001264734.1 | 9 - 10 | 966 | 69 | ||
| NM_008995.2 | NP_033021.2 | 9 - 10 | 929 | 69 | ||
| NM_175933.2 | NP_787947.1 | 8 - 9 | 855 | 69 | ||
| GenBank mRNA | AJ416473.1 | - | 8 - 9 | 837 | 69 | |
| AK088886.1 | - | 8 - 9 | 859 | 69 | ||
| AK145111.1 | - | 9 - 10 | 902 | 69 | ||
| AK145361.1 | - | 8 - 9 | 792 | 69 | ||
| AK161470.1 | - | 8 - 9 | 852 | 69 | ||
| BC029748.1 | - | 9 - 10 | 936 | 2599159 | 69 | |
| Z97018.1 | - | 8 - 9 | 854 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex5
Genomic Map