Assay Details
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013700.1 | NP_038728.1 | 7 - 8 | 921 | 74 | |
| GenBank mRNA | AK154008.1 | - | 7 - 8 | 910 | 74 | |
| BC066993.1 | - | 7 - 8 | 899 | 5689229 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp5
Genomic Map