Assay Details
| Gene Symbol: Cd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013488.2 | NP_038516.1 | 6 - 7 | 1126 | 101 | |
| GenBank mRNA | AF045882.1 | - | 4 - 5 | 885 | 101 | |
| AK040115.1 | - | 6 - 7 | 1118 | 101 | ||
| AK153966.1 | - | 6 - 7 | 1126 | 101 | ||
| AK161775.1 | - | 6 - 7 | 1050 | 101 | ||
| BC039137.1 | - | 6 - 7 | 1105 | 3670597 | 101 | |
| M13816.1 | - | 5 - 6 | 996 | 101 | ||
| M36850.1 | - | 5 - 6 | 996 | 101 | ||
| U75219.1 | - | 1 - 2 | 30 | 101 | ||
| X04836.1 | - | 6 - 7 | 1126 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd4
Genomic Map