Assay Details
| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081047.1 | NP_001074516.1 | 9 - 10 | 943 | 72 | |
| GenBank mRNA | AK145910.1 | - | 9 - 10 | 72 | ||
| BC113805.1 | - | 9 - 10 | 908 | 40062208 | 72 | |
| BC113956.1 | - | 9 - 10 | 908 | 40062209 | 72 | |
More Information
Set Membership: |
|

Cnksr1
Genomic Map