Assay Details
| Gene Symbol: Kcnq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081142.1 | NP_001074611.1 | 6 - 7 | 954 | 66 | |
| GenBank mRNA | AK040190.1 | - | 6 - 7 | 887 | 66 | |
More Information
Set Membership: |
|

Kcnq4
Genomic Map