Assay Details
| Gene Symbol: Kcnq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081142.1 | NP_001074611.1 | 8 - 9 | 1136 | 76 | |
| GenBank mRNA | AK040190.1 | - | 8 - 9 | 1069 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnq4
Genomic Map