Assay Details
| Gene Symbol: Mettl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177793.3 | NP_808461.1 | 1 - 2 | 386 | 58 | |
| GenBank mRNA | AK033488.1 | - | 1 - 2 | 387 | 58 | |
| BC147316.1 | - | 1 - 2 | 352 | 9056371 | 58 | |
| BC147317.1 | - | 1 - 2 | 352 | 9056368 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl24
Genomic Map