Assay Details
| Gene Symbol: Nr2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152229.2 | NP_689415.1 | 3 - 4 | 940 | 92 | |
| GenBank mRNA | AK132201.1 | - | 3 - 4 | 944 | 92 | |
| BC056950.1 | - | 3 - 4 | 571 | 92 | ||
| BC057104.1 | - | 3 - 4 | 984 | 6401770 | 92 | |
| S77482.1 | - | 3 - 4 | 983 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr2e1
Genomic Map