Assay Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168333.1 | NP_001161805.1 | 7 - 8 | 940 | 60 | |
| NM_023476.3 | NP_075965.2 | 7 - 8 | 1081 | 60 | ||
| GenBank mRNA | AB050626.1 | - | 7 - 8 | 918 | 60 | |
| AK087142.1 | - | 941 | 60 | |||
| BC018539.1 | - | 7 - 8 | 925 | 3596560 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tinagl1
Genomic Map