Assay Details
| Gene Symbol: A430078I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040219.1 | - | 105 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | expressed sequence C87436 | C87436 | -2.4kb | Anti-sense | Chr. 6 - 86437328 - 86470388 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2310040G24 gene | 2310040G24Rik | 2.6kb | Sense | Chr. 6 - 86483376 - 86488227 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

A430078I02Rik
Genomic Map